siRNA Details
Gene Symbol: Gpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122963.1 | NP_001116435.1 | 10 | 2339 | ||
NM_028487.4 | NP_082763.3 | 11 | 2399 | |||
GenBank mRNA | AK135728.1 | - | 11 | 2330 | ||
AK136894.1 | - | 11 | 2179 | |||
AK145318.1 | - | 10 | 2328 | |||
AK151283.1 | - | 9 | 1560 | |||
AK163370.1 | - | 11 | 2247 | |||
AK163390.1 | - | 11 | 2401 | |||
AK169406.1 | - | 11 | 2194 | |||
AY382529.1 | - | 11 | 2197 | |||
BC016083.1 | - | 9 | 1193 | 4501399 | ||
BC067075.1 | - | 10 | 2113 | 30614988 | ||
BC076635.1 | - | 10 | 2169 | |||
BC094888.1 | - | 10 | 2132 | 30628044 | ||
BC096442.1 | - | 11 | 2214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
