siRNA Details
Gene Symbol: Lce1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028628.2 | NP_082904.1 | 2 | 506 | ||
GenBank mRNA | AK003585.1 | - | 2 | 507 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lce1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028628.2 | NP_082904.1 | 2 | 506 | ||
GenBank mRNA | AK003585.1 | - | 2 | 507 |