siRNA Details
Gene Symbol: 1110034G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028637.1 | NP_082913.1 | 3 | 881 | ||
GenBank mRNA | AK004090.1 | - | 3 | 455 | ||
BC100475.1 | - | 3 | 881 | 30253755 | ||
BC138171.1 | - | 3 | 535 | 8861190 |
