siRNA Details
Gene Symbol: 2810403A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028814.3 | NP_083090.2 | 7 | 856 | ||
GenBank mRNA | AK012969.1 | - | 7 | 833 | ||
AK083018.1 | - | 7 | 817 | |||
AK087126.1 | - | 1 | 524 | |||
AK170490.1 | - | 7 | 851 | |||
AK173057.1 | - | 7 | 820 | |||
AK193095.1 | - | 1 | 70 | |||
BC006621.1 | - | 7 | 831 | 3158104 |
