siRNA Details
Gene Symbol: 1700080O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028851.1 | NP_083127.1 | 2 | 109 | ||
GenBank mRNA | AK006959.1 | - | 2 | 110 | ||
BC109337.2 | - | 1 | 39 | 40052428 | ||
BC109338.1 | - | 1 | 39 | 40052437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
