siRNA Details
Gene Symbol: 4931414P19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028890.2 | NP_083166.1 | 7 | 1837 | ||
GenBank mRNA | AK016457.1 | - | 6 | 1310 | ||
BC029624.1 | - | 7 | 1837 | 3992239 | ||
BC034212.1 | - | 7 | 1838 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
