siRNA Details
Gene Symbol: Abca12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175210.3 | NP_780419.2 | 2 | 411 | ||
GenBank mRNA | AK014713.1 | - | 2 | 399 | ||
AK028781.1 | - | 2 | 412 | |||
AK029031.1 | - | 2 | 412 | |||
BC158053.1 | - | 2 | 224 | 9088162 | ||
BC158063.1 | - | 2 | 224 | 9007362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
