siRNA Details
Gene Symbol: 4930481A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015604.1 | - | 4 | 608 | ||
AK039982.1 | - | 4 | 594 | |||
BC065793.1 | - | 4 | 537 | |||
BC085508.1 | - | 4 | 548 | |||
BM937611.1 | - | 3 | 431 |
