siRNA Details
Gene Symbol: Ndufaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127346.1 | NP_001120818.1 | 1, 2 | 192 | ||
GenBank mRNA | AB183434.1 | - | 1, 2 | 136 | ||
AK007894.1 | - | 1, 2 | 183 | |||
AK166492.1 | - | 1, 2 | 159 | |||
AY262012.1 | - | 1, 2 | 164 | |||
BC048512.1 | - | 1, 2 | 151 | |||
BC116975.1 | - | 1, 2 | 158 | 40126294 | ||
BC116977.1 | - | 1, 2 | 158 | 40126296 | ||
BC144716.1 | - | 1, 2 | 158 | |||
BY251841.1 | - | 1, 2 | 193 |
