siRNA Details
Gene Symbol: 1700020A23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163483.1 | NP_001156955.1 | 2 | 661 | ||
NM_029375.1 | NP_083651.1 | 3 | 484 | |||
GenBank mRNA | AK006141.1 | - | 2 | 466 | ||
BC049710.1 | - | 2 | 661 |
