siRNA Details
Gene Symbol: Tex22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029381.1 | NP_083657.1 | 3 | 1190 | ||
GenBank mRNA | AF522069.1 | - | 3 | 1190 | ||
AK006471.1 | - | 3 | 658 | |||
BC048634.1 | - | 3 | 623 | 6770610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
