siRNA Details
Gene Symbol: Ccdc125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168386.2 | NP_001161858.1 | 5, 6 | 911 | ||
NM_183115.5 | NP_898938.4 | 6, 7 | 989 | |||
GenBank mRNA | AK017973.1 | - | 5, 6 | 674 | ||
BC085249.1 | - | 6, 7 | 707 | 30726150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
