siRNA Details
Gene Symbol: Ccdc125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168386.2 | NP_001161858.1 | 2 | 592 | ||
NM_183115.5 | NP_898938.4 | 2 | 592 | |||
GenBank mRNA | AK017973.1 | - | 2 | 355 | ||
BC085249.1 | - | 2 | 310 | 30726150 | ||
BU513278.1 | - | 3 | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
