siRNA Details
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029512.2 | NP_083788.2 | 4 | 787 | ||
NM_181734.3 | NP_859423.2 | 4 | 784 | |||
GenBank mRNA | AK018052.1 | - | 3 | 554 | ||
AK033600.1 | - | 4 | 733 | |||
AK039450.1 | - | 4 | 785 | |||
AK043791.1 | - | 4 | 754 | |||
AK081680.1 | - | 4 | 747 | |||
AK156641.1 | - | 4 | 750 | |||
BC052389.1 | - | 4 | 743 | 5718874 | ||
BC060726.1 | - | 3 | 653 | |||
BY717985.1 | - | 3 | 554 | |||
CK640285.1 | - | 3 | 402 |

More Information
Other Species Targeted

Gene Ontology Categories:
