siRNA Details
Gene Symbol: Adhfe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175236.4 | NP_780445.1 | 13 | 1340 | ||
GenBank mRNA | AK018241.1 | - | 13 | 1311 | ||
AK038853.1 | - | 13 | 1307 | |||
AK046780.1 | - | 13 | 1266 | |||
AK050178.1 | - | 12 | 1264 | |||
AK084104.1 | - | 1269 | ||||
BC026584.1 | - | 13 | 1298 | 4972323 |
