siRNA Details
Gene Symbol: 1700112E06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028275.1 | NP_082551.1 | 3, 4 | 423 | ||
GenBank mRNA | AK007175.1 | - | 3, 4 | 427 | ||
AK012226.1 | - | 1, 2 | 248 | |||
AK153491.1 | - | 2, 3 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
