siRNA Details
Gene Symbol: Kdm4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172095.1 | NP_001165566.1 | 18 | 2721 | ||
NM_144787.2 | NP_659036.1 | 18 | 2785 | |||
GenBank mRNA | AK144103.1 | - | 18 | 2748 | ||
AK173023.1 | - | 18 | 2785 | |||
BC020180.1 | - | 18 | 2727 | 4923943 | ||
BC042424.1 | - | 18 | 2721 | 5357211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
