siRNA Details
Gene Symbol: Kdm4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172095.1 | NP_001165566.1 | 4 | 511 | ||
NM_144787.2 | NP_659036.1 | 4 | 575 | |||
GenBank mRNA | AK042792.1 | - | 3 | 290 | ||
AK048092.1 | - | 4 | 488 | |||
AK053104.1 | - | 4 | 451 | |||
AK081769.1 | - | 4 | 518 | |||
AK144103.1 | - | 4 | 538 | |||
AK144890.1 | - | 4 | 549 | |||
AK173023.1 | - | 4 | 575 | |||
BC020180.1 | - | 4 | 517 | 4923943 | ||
BC042424.1 | - | 4 | 511 | 5357211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
