siRNA Details
Gene Symbol: Calcoco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271018.1 | NP_001257947.1 | 3, 4 | 304 | ||
GenBank mRNA | AF490344.1 | - | 3, 4 | 302 | ||
AK010816.1 | - | 3, 4 | 302 | |||
AK145542.1 | - | 3, 4 | 305 | |||
AK166710.1 | - | 3, 4 | 305 | |||
AK166756.1 | - | 3, 4 | 305 | |||
AK166939.1 | - | 3, 4 | 302 | |||
AK167009.1 | - | 3, 4 | 305 | |||
AK167172.1 | - | 3, 4 | 305 | |||
AK167291.1 | - | 3, 4 | 304 |
