siRNA Details
Gene Symbol: Mrap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029844.3 | NP_084120.1 | 3 | 943 | ||
GenBank mRNA | AK003912.2 | - | 3 | 944 | ||
AK158113.1 | - | 3 | 817 | |||
AY079153.1 | - | 3 | 720 | |||
BC027543.1 | - | 3 | 797 | 1477097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
