siRNA Details
Gene Symbol: 2210407C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144544.2 | NP_653127.2 | 3 | 195 | ||
GenBank mRNA | AA124355.1 | - | 3 | 182 | ||
AY148487.1 | - | 2 | 101 | |||
BC019553.1 | - | 3 | 173 | 4456658 | ||
BY096725.1 | - | 3 | 188 | |||
BY097425.1 | - | 3 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
