siRNA Details
Gene Symbol: Cypt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177380.1 | NP_001170851.1 | 2 | 222 | ||
GenBank mRNA | AK018908.1 | - | 2 | 222 | ||
BC100466.1 | - | 2 | 222 |
Gene Symbol: Cypt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191032.1 | NP_001177961.1 | 2 | 222 | ||
GenBank mRNA | BC099555.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
