siRNA Details
Gene Symbol: Bola3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175277.4 | NP_780486.1 | 2 | 102 | ||
GenBank mRNA | AK028102.1 | - | 2 | 103 | ||
AK131950.1 | - | 111 | ||||
AK160488.1 | - | 2 | 106 | |||
AK213244.1 | - | 1 | 37 | |||
BC049748.1 | - | 2 | 122 | |||
BC116947.1 | - | 2 | 140 | 40126266 | ||
BC116949.1 | - | 2 | 140 | 40126268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
