siRNA Details
Gene Symbol: Zc3hav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028421.1 | NP_082697.1 | 4, 5 | 2124 | ||
NM_028864.2 | NP_083140.1 | 4, 5 | 2124 | |||
GenBank mRNA | AK004770.1 | - | 4, 5 | 2125 | ||
AK020263.2 | - | 4, 5 | 1803 | |||
AK080334.1 | - | 4, 5 | 2109 | |||
AK143568.1 | - | 4, 5 | 1794 | |||
AK169402.1 | - | 4, 5 | 1769 | |||
AK172379.1 | - | 4, 5 | 2125 |
