siRNA Details
Gene Symbol: Lin28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145833.1 | NP_665832.1 | 4 | 1134 | ||
GenBank mRNA | AF521097.1 | - | 4 | 1134 | ||
BC068304.1 | - | 4 | 1091 | 30432006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
