siRNA Details
Gene Symbol: Rnf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033604.2 | NP_291082.1 | 14 | 3088 | ||
GenBank mRNA | AF330197.1 | - | 14 | 3084 | ||
AK164502.1 | - | 5 | 485 | |||
BC054842.1 | - | 4 | 327 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106836, XM_006243372, XM_008766243, XM_006243377, XM_008766244, and others.
Gene Ontology Categories:
