siRNA Details
Gene Symbol: Klra21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053151.1 | NP_444381.1 | 305 | |||
GenBank mRNA | AF288380.1 | - | 305 | |||
AY338977.1 | - | 262 | ||||
AY338978.1 | - | 240 | ||||
AY860976.1 | - | 2 | 211 | |||
AY971806.1 | - | 199 | ||||
BC064073.1 | - | 308 | 874161 | |||
BC116399.1 | - | 273 | 40101978 | |||
BC116400.1 | - | 283 | 40101979 | |||
FJ159529.1 | - | 193 |
Gene Symbol: Klra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC141217.1 | - | 289 | 9055829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
