siRNA Details
Gene Symbol: Med15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040683.2 | NP_001035773.1 | 12 | 1697 | ||
NM_001285884.1 | NP_001272813.1 | 13 | 1817 | |||
NM_001285886.1 | NP_001272815.1 | 12 | 1509 | |||
NM_033609.3 | NP_291087.2 | 13 | 1817 | |||
GenBank mRNA | AF328770.1 | - | 13 | 1693 | ||
AK131186.1 | - | 12 | 1638 | |||
AK142224.1 | - | 13 | 1784 | |||
AK166522.1 | - | 12 | 1509 | |||
AK169904.1 | - | 13 | 1818 | |||
BC003221.1 | - | 3 | 256 | |||
BC049891.1 | - | 8 | 1281 | |||
BC054779.1 | - | 12 | 1647 | 6812541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
