siRNA Details
Gene Symbol: Wwtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168281.1 | NP_001161753.1 | 3, 4 | 766 | ||
NM_133784.3 | NP_598545.2 | 3, 4 | 845 | |||
GenBank mRNA | AJ299430.1 | - | 3, 4 | 780 | ||
AK009983.1 | - | 3, 4 | 778 | |||
AK011504.1 | - | 3, 4 | 820 | |||
AK142299.1 | - | 3, 4 | 767 | |||
AK142720.1 | - | 3, 4 | 770 | |||
BC004640.1 | - | 3, 4 | 766 | 3157246 | ||
BC014727.1 | - | 3, 4 | 797 | 4487316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
