siRNA Details
Gene Symbol: C87977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177542.1 | NP_001171013.1 | 3 | 439 | ||
GenBank mRNA | AK139502.1 | - | 3 | 438 | ||
BC094895.1 | - | 3 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
