siRNA Details
Gene Symbol: B3gnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271915.1 | NP_001258844.1 | 1 | 1337 | ||
NM_178879.4 | NP_849210.1 | 2 | 1149 | |||
GenBank mRNA | AK032108.1 | - | 1 | 1338 | ||
AK053867.1 | - | 1 | 851 | |||
AK169360.1 | - | 1 | 1333 | |||
BC047209.1 | - | 1 | 1309 | |||
BC108347.1 | - | 1 | 1287 | 5010813 | ||
BC120583.1 | - | 1 | 1002 | 40129506 | ||
BC120585.1 | - | 1 | 1002 | 40129508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
