siRNA Details
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098520.1 | NP_001091990.1 | 5 | 653 | ||
NM_001098521.1 | NP_001091991.1 | 5 | 581 | |||
NM_001098522.1 | NP_001091992.1 | 4 | 863 | |||
NM_006410.4 | NP_006401.3 | 5 | 548 | |||
GenBank mRNA | AF039103.1 | - | 4 | 494 | ||
AK222969.1 | - | 4 | 537 | |||
AK223010.1 | - | 4 | 571 | |||
AK292092.1 | - | 5 | 557 | |||
BC002439.2 | - | 5 | 594 | 3346984 | ||
BC015358.2 | - | 4 | 559 | 3455757 | ||
U69161.1 | - | 4 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
