siRNA Details
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098520.1 | NP_001091990.1 | 4 | 559 | ||
NM_001098521.1 | NP_001091991.1 | 4 | 487 | |||
NM_001098522.1 | NP_001091992.1 | 3 | 769 | |||
NM_006410.4 | NP_006401.3 | 4 | 454 | |||
GenBank mRNA | AF039103.1 | - | 3 | 400 | ||
AK222969.1 | - | 3 | 443 | |||
AK223010.1 | - | 3 | 477 | |||
AK292092.1 | - | 4 | 463 | |||
BC002439.2 | - | 4 | 500 | 3346984 | ||
BC015358.2 | - | 3 | 465 | 3455757 | ||
DB450790.1 | - | 4 | 485 | |||
U69161.1 | - | 3 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
