siRNA Details
Gene Symbol: PTTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006607.2 | NP_006598.2 | 1 | 216 | ||
GenBank mRNA | AF095288.1 | - | 1 | 216 | ||
BC069114.1 | - | 1 | 216 | 7216932 | ||
BC069400.1 | - | 1 | 216 | 7262134 |
Gene Symbol: PTTG3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002734.2 | 1 | 245 | |||
GenBank mRNA | AF095289.1 | - | 1 | 216 |
