siRNA Details
Gene Symbol: Mfap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026220.4 | NP_080496.1 | 1, 2 | 195 | ||
GenBank mRNA | AK012688.1 | - | 1, 2 | 181 | ||
AK014483.1 | - | 1, 2 | 188 | |||
AK032586.1 | - | 1, 2 | 196 | |||
AK050561.1 | - | 1, 2 | 196 | |||
AK077604.1 | - | 1, 2 | 189 | |||
AK088766.1 | - | 1, 2 | 192 | |||
AK161000.1 | - | 1, 2 | 196 | |||
AK168801.1 | - | 1, 2 | 201 |
Gene Symbol: Mfap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081975.3 | NP_001075444.1 | 1, 2 | 191 | ||
GenBank mRNA | AK011317.1 | - | 1, 2 | 189 | ||
AK018514.1 | - | 1, 2 | 192 | |||
AK088946.1 | - | 198 | ||||
AK169029.1 | - | 199 | ||||
BC005728.1 | - | 1, 2 | 152 | 3595848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
