siRNA Details
Gene Symbol: Mfap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026220.4 | NP_080496.1 | 3 | 772 | ||
GenBank mRNA | AK014483.1 | - | 3 | 765 | ||
AK032586.1 | - | 3 | 773 | |||
AK050561.1 | - | 3 | 773 | |||
AK077604.1 | - | 3 | 766 | |||
AK161000.1 | - | 3 | 773 | |||
AK168801.1 | - | 3 | 778 |
Gene Symbol: Mfap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081975.3 | NP_001075444.1 | 3 | 768 | ||
GenBank mRNA | AK018514.1 | - | 3 | 769 | ||
AK169029.1 | - | 776 | ||||
AK178130.1 | - | 2 | 386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
