siRNA Details
Gene Symbol: Nudt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031664.1 | NP_001026834.1 | 2 | 1327 | ||
GenBank mRNA | BC055771.1 | - | 2 | 1284 | 6404994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
