siRNA Details
Gene Symbol: Zfp119a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144546.6 | NP_653129.1 | 4 | 857 | ||
GenBank mRNA | AF242376.1 | - | 4 | 858 | ||
BC118950.2 | - | 4 | 834 | 40092426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
