siRNA Details
Gene Symbol: Npffr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133192.3 | NP_573455.2 | 4 | 804 | ||
GenBank mRNA | AF236084.1 | - | 3 | 698 | ||
AF330054.1 | - | 3 | 689 | |||
AK050939.1 | - | 4 | 805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
