siRNA Details
Gene Symbol: AW209491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104646.1 | NP_001098116.1 | 2 | 596 | ||
NM_134067.4 | NP_598828.3 | 2 | 663 | |||
GenBank mRNA | AK030506.1 | - | 645 | |||
AK050507.1 | - | 2 | 645 | |||
AK077649.1 | - | 2 | 645 | |||
AK167401.1 | - | 2 | 664 | |||
AK169400.1 | - | 2 | 596 | |||
BC016098.1 | - | 1 | 560 | 4910571 | ||
BC034309.1 | - | 1 | 419 | 5362794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
