siRNA Details
Gene Symbol: Akr1c14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134072.1 | NP_598833.1 | 3 | 310 | ||
GenBank mRNA | AK033537.1 | - | 3 | 328 | ||
AK136528.1 | - | 3 | 327 | |||
AY730283.1 | - | 3 | 272 | |||
BC013482.1 | - | 3 | 310 | 4240772 | ||
BC026628.1 | - | 3 | 309 | 4983237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
