siRNA Details
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178920.4 | NP_849251.1 | 1 | 246 | ||
GenBank mRNA | AK035986.1 | - | 1 | 128 | ||
BC113154.1 | - | 1 | 123 | 40058489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
