siRNA Details
Gene Symbol: Topors | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134097.3 | NP_598858.2 | 3 | 2753 | ||
GenBank mRNA | AB072395.1 | - | 3 | 2710 | ||
AB104865.1 | - | 3 | 2738 | |||
BC019421.1 | - | 3 | 2711 | |||
BC037141.1 | - | 3 | 2731 | |||
BC040797.1 | - | 3 | 2741 | 3492178 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108658.
Gene Ontology Categories:
