siRNA Details
Gene Symbol: Topors | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134097.3 | NP_598858.2 | 3 | 2224 | ||
GenBank mRNA | AB072395.1 | - | 3 | 2181 | ||
AB104865.1 | - | 3 | 2209 | |||
AK134075.1 | - | 3 | 2220 | |||
AK143025.1 | - | 3 | 2205 | |||
AK153743.1 | - | 3 | 2211 | |||
BC019421.1 | - | 3 | 2182 | |||
BC037141.1 | - | 3 | 2202 | |||
BC040797.1 | - | 3 | 2212 | 3492178 |

More Information
Other Species Targeted

Gene Ontology Categories:
