siRNA Details
Gene Symbol: Topors | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134097.3 | NP_598858.2 | 3 | 982 | ||
GenBank mRNA | AB072395.1 | - | 3 | 939 | ||
AB104865.1 | - | 3 | 967 | |||
AK044564.1 | - | 3 | 960 | |||
AK134075.1 | - | 3 | 978 | |||
AK140250.1 | - | 3 | 983 | |||
AK143025.1 | - | 3 | 963 | |||
AK153743.1 | - | 3 | 969 | |||
BC019421.1 | - | 3 | 940 | |||
BC037141.1 | - | 3 | 960 | |||
BC040797.1 | - | 3 | 970 | 3492178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
