siRNA Details
Gene Symbol: Exoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175353.2 | NP_780562.2 | 19 | 2081 | ||
GenBank mRNA | AK076455.1 | - | 2 | 254 | ||
AK151524.1 | - | 19 | 2082 | |||
AK151838.1 | - | 19 | 2082 | |||
BC025616.1 | - | 6 | 655 | |||
BC026859.1 | - | 19 | 2034 | 2631201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
