siRNA Details
Gene Symbol: Gtpbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145147.5 | NP_660129.2 | 10 | 1526 | ||
GenBank mRNA | AJ293619.1 | - | 9 | 1156 | ||
AK082192.1 | - | 7 | 1402 | |||
AK144812.1 | - | 10 | 1527 | |||
AK152225.1 | - | 10 | 1527 | |||
AK152972.1 | - | 10 | 1527 | |||
BC027821.1 | - | 7 | 1057 | 5321725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
