siRNA Details
Gene Symbol: Ints5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176843.3 | NP_789813.1 | 2 | 1031 | ||
GenBank mRNA | AK143868.1 | - | 2 | 994 | ||
AK144770.1 | - | 2 | 1006 | |||
AK155499.1 | - | 2 | 993 | |||
AK166149.1 | - | 2 | 1004 | |||
AK168795.1 | - | 2 | 1004 | |||
AK170584.1 | - | 2 | 993 | |||
AK172260.1 | - | 2 | 993 | |||
BC039214.1 | - | 2 | 1013 | 4191047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
