siRNA Details
Gene Symbol: Dut | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159646.1 | NP_001153118.1 | 4 | 413 | ||
NM_023595.6 | NP_076084.2 | 3 | 508 | |||
GenBank mRNA | AF004565.2 | - | 312 | |||
AF091101.1 | - | 3 | 365 | |||
AK003076.1 | - | 3 | 297 | |||
AK011407.1 | - | 3 | 299 | |||
AK017216.1 | - | 4 | 403 | |||
AK077244.1 | - | 3 | 541 | |||
AK135438.1 | - | 4 | 422 | |||
AK152459.1 | - | 3 | 304 | |||
AK153285.1 | - | 3 | 289 | |||
BC019979.1 | - | 4 | 395 | 4187559 | ||
BC053693.1 | - | 3 | 295 | 6485182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
