siRNA Details
Gene Symbol: Nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083906.1 | NP_001077375.1 | 2 | 1380 | ||
GenBank mRNA | BC133713.1 | - | 2 | 1380 | 40143429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
